Få mere at vide
Testosterone Propionate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Testosterone propionate |
| CAS | 57-85-2 |
| Molekylær formel | C22 H32 O3 |
| Mængde | 5 g |
| Synonym | Testosterone Propionate, Testosterone Isocaproate Imp. C (EP), Androst-4-en-3-one','17-(1-oxopropoxy)-','(17β)-, Testosterone','propionate (8CI), (17β)-17-(1-Oxopropoxy)androst-4-en-3-one, 17β-(Propionyloxy)androst-4-en-3-one, 17β-Hydroxy-4-androsten-3-one 17-propionate, 17β-Hydroxyandrost-4-en-3-one propionate, 17β-Hydroxyandrost-4-en-3-one-17β-propionate, Agovirin, Androlon, Androtest P, Androteston, Anertan, Aquaviron, Bio-testiculina, Enarmon, Enarmon-oil, Homandren, Hormoteston, Masenate, NRB 03689, NSC 9166, Nasdol, Okasa-Mascul, Orchiol, Orchisterone, Orchistin, Oreton, Pantestin, Primotestone, Propiokan, Recthormone testosterone, Solvotest, Sterandryl, Synandrol, Synerone, TP, Telipex, Testaform, Testex, Testodet, Testodrin, Testogen, Testolets, Testonique, Testormol, Testosid, Testosteron propionate, Testosterone-17-propionate, Testosterone-17β propionate, Testoviron, Testoviron (ampule), Testoxyl, Testrex, Textes Depot, Tostrin, Uniteston, Virormone, neo-Hombreol |
| InChI formel | InChI=1S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1 |
| SMIL | CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC navn | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] propanoate |
| Molekylvægt (g/mol) | 344.49 |
| Formel vægt | 344.2351 |