Få mere at vide
Testosteron Enanthat, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Testosterone enanthate |
| CAS | 315-37-7 |
| Molekylær formel | C26 H40 O3 |
| Mængde | 25 mg |
| Synonym | Testosterone Enantate, Testosterone Isocaproate Imp. F (EP), Androst-4-en-3-one','17-[(1-oxoheptyl)oxy]-','(17β)-, Testosterone','heptanoate (6CI,8CI), (17β)-17-[(1-Oxoheptyl)oxy]androst-4-en-3-one, Heptanoic acid','ester with testosterone (6CI), 17β-Enanthoxyandrost-4-en-3-one, 17β-Hydroxyandrost-4-en-3-one enanthate, 4-Androsten-3-one 17β-enanthate, Andro LA 200, Androtardyl, Delatestryl, Enarmone depot, Everone, Jenasteron, NSC 17591, Primoteston-Depot, Reposo TMD, Testenate, Testinon, Testo-Enant, Testosterone 17-enanthate, Testosterone enanthate, Testosterone heptylate, Testosterone oenanthate |
| InChI formel | InChI=1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1 |
| SMIL | CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC navn | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoat |
| Molekylvægt (g/mol) | 400.59 |
| Formel vægt | 400.2977 |