Få mere at vide
Testosterone-2,2,4,6,6-d5, CDN
Artikelnummer. 30321103
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321103 0.01 g
30318663 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321103 Leverandør CDN Leverandørnr. CDND59170.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Testosterone-2,2,4,6,6-d5
CAS 21002-80-2
Molekylær formel C19 2H5 H23 O2
Mængde 0.01 g
Synonym Androst-4-en-3-one-2,2,4,6,6-d5, 17-hydroxy-, (17β)-, Testosterone-2,2,4,6,6-d5 (8 CI), (17β)-17-Hydroxyandrost-4-en-3-one-2,2,4,6,6-d5
InChI formel InChI=1 S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h11,14-17,21 H,3-10H2,1-2H3/t14-,15-,16-,17-,18-,19-/m0/s1/i3D2,7D2,11 D
SMIL [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H](O)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O
IUPAC navn (8 R,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6-pentadeuterio-17-hydroxy-10,13-dimethyl-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
Molekylvægt (g/mol) 293.46
Formel vægt 293.2403 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity