Få mere at vide
Testosterone 17-propionate, Lipomed
Artikelnummer. 30458635
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30458635 10 mg
1 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30458635 Leverandør Lipomed Leverandørnr. LPMTES124010
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Testosterone 17-propionate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Testosterone propionate
CAS 57-85-2
Analyt- eller komponentnavne Testosterone propionate
Forsendelsestilstand Room Temperature
Molekylær formel C22 H32 O3
Synonym Testosterone Isocaproate Imp. C (EP), Testosterone Propionate, Androst-4-en-3-one, 17-(1-oxopropoxy)-, (17 beta)-, Testosterone, propionate (8 CI), (17 beta)-17-(1-Oxopropoxy)androst-4-en-3-one, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMIL CCC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C
IUPAC navn [(8 R,9 S,10 R,13 S,14 S,17 S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] propanoate
Industry Type Pharmaceutical
Molekylvægt (g/mol) 344.49
Mængde 10 mg
Formel vægt 344.2351 g/mol