Få mere at vide
Testosterone 17-propionate, Lipomed
Beskrivelse
Testosterone 17-propionate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Testosterone propionate |
| CAS | 57-85-2 |
| Analyt- eller komponentnavne | Testosterone propionate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C22 H32 O3 |
| Synonym | Testosterone Isocaproate Imp. C (EP), Testosterone Propionate, Androst-4-en-3-one, 17-(1-oxopropoxy)-, (17 beta)-, Testosterone, propionate (8 CI), (17 beta)-17-(1-Oxopropoxy)androst-4-en-3-one, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate, Testosterone-17 beta propionate |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMIL | CCC(=O)O[C@H]1 CC[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC navn | [(8 R,9 S,10 R,13 S,14 S,17 S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] propanoate |
| Industry Type | Pharmaceutical |
| Vis mere |