missing translation for 'onlineSavingsMsg'
Få mere at vide

Testosterone 17-Isocaproate, TRC

Artikelnummer. 30418979
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418979 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30418979 Leverandør Toronto Research Chemicals Leverandørnr. TRCT15508510MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Testosterone isocaproate
CAS 15262-86-9
Molekylær formel C25 H38 O3
Mængde 10 mg
Synonym Androst-4-en-3-one','17-[(4-methyl-1-oxopentyl)oxy]-','(17β)-, Testosterone','4-methylvalerate (6CI,8CI), (17β)-17-[(4-Methyl-1-oxopentyl)oxy]androst-4-en-3-one, Valeric acid','4-methyl-','ester with testosterone (8CI), 3-Oxoandrost-4-en-17β-yl isocaproate, NSC 26641, Testosterone 17-isocaproate, Testosterone 4-methylpentanoate, Testosterone isocaproate
InChI formel InChI=1S/C25H38O3/c1-16(2)5-10-23(27)28-22-9-8-20-19-7-6-17-15-18(26)11-13-24(17,3)21(19)12-14-25(20,22)4/h15-16,19-22H,5-14H2,1-4H3/t19-,20-,21-,22-,24-,25-/m0/s1
SMIL CC(C)CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC navn [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 4-methylpentanoate
Molekylvægt (g/mol) 386.57
Formel vægt 386.2821
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.