Learn More
Testosterone 17-Isocaproate, TRC
Product Code. 30422628
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
30422628 100 mg 100mg
1 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 30422628 Supplier Toronto Research Chemicals Supplier No. TRCT155085100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Testosterone isocaproate
CAS 15262-86-9
Molecular Formula C25 H38 O3
Quantity 100 mg
Synonym Androst-4-en-3-one','17-[(4-methyl-1-oxopentyl)oxy]-','(17β)-, Testosterone','4-methylvalerate (6CI,8CI), (17β)-17-[(4-Methyl-1-oxopentyl)oxy]androst-4-en-3-one, Valeric acid','4-methyl-','ester with testosterone (8CI), 3-Oxoandrost-4-en-17β-yl isocaproate, NSC 26641, Testosterone 17-isocaproate, Testosterone 4-methylpentanoate, Testosterone isocaproate
InChI Formula InChI=1S/C25H38O3/c1-16(2)5-10-23(27)28-22-9-8-20-19-7-6-17-15-18(26)11-13-24(17,3)21(19)12-14-25(20,22)4/h15-16,19-22H,5-14H2,1-4H3/t19-,20-,21-,22-,24-,25-/m0/s1
SMILES CC(C)CCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] 4-methylpentanoate
Molecular Weight (g/mol) 386.57
Formula Weight 386.2821