Få mere at vide
Testosterone 17-Hexahydrobenzoate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Testosterone 17-Hexahydrobenzoate |
| CAS | 14191-92-5 |
| Molekylær formel | C26 H38 O3 |
| Mængde | 10 mg |
| Synonym | Sterandryl-A, Tardosterandryl-Depot, Testodur, Testosterone 17-Hexahydrobenzoate, Testosterone Cyclohexanoate, Testosterone Hexahydrobenzoate, Virex-Retard, Benzotest, Durandron, Sterandryl Retard, Cyclohexanecarboxylate Testosterone, Cyclohexanecarboxylic Acid Ester with Testosterone |
| InChI formel | InChI=1S/C26H38O3/c1-25-14-12-19(27)16-18(25)8-9-20-21-10-11-23(26(21,2)15-13-22(20)25)29-24(28)17-6-4-3-5-7-17/h16-17,20-23H,3-15H2,1-2H3/t20?,21?,22?,23-,25-,26-/m0/s1 |
| SMIL | C[C@]12CCC3C(CCC4=CC(=O)CC[C@]34C)C1CC[C@@H]2OC(=O)C5CCCCC5 |
| IUPAC navn | [(10R,13S,17S)-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] cyclohexanecarboxylate |
| Molekylvægt (g/mol) | 398.58 |
| Formel vægt | 398.2821 |