missing translation for 'onlineSavingsMsg'
Få mere at vide

Testosteron-16,16,17-d3 propionat, CDN

Artikelnummer. 30325540
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30325540 0.005 g
30326538 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30325540 Leverandør CDN Leverandørnr. CDND72120.005G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Opdag 4000+ højkvalitets deutererede forbindelser, der sendes direkte fra lager for hurtighed. Vi tilbyder mange unikke API'er, herunder aktive farmaceutiske ingredienser, nitrosaminer, aminosyrer, steroider, gasser, pesticider og fedtsyrer.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Testosterone-16,16,17-d3 Propionate
CAS 876054-54-5
Molekylær formel C22 2H3 H29 O3
Mængde 0.005 g
Synonym Androst-4-en-3-one-16,16,17-d3, 17-(1-oxopropoxy)-, (17β)- (9 CI), Testosterone-d3 propionate, Testosterone-16,16,17-d3 propionate
InChI formel InChI=1 S/C22H32O3/c1-4-20(24)25-19-8-7-17-16-6-5-14-13-15(23)9-11-21(14,2)18(16)10-12-22(17,19)3/h13,16-19 H,4-12H2,1-3H3/t16-,17-,18-,19-,21-,22-/m0/s1/i8D2,19 D
SMIL [2 H]C1([2 H])C[C@H]2 [C@@H]3 CCC4=CC(=O)CC[C@]4(C)[C@H]3 CC[C@]2(C)[C@@]1([2 H])OC(=O)CC
IUPAC navn [(8 R,9 S,10 R,13 S,14 S,17 S)-16,16,17-trideuterio-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15-decahydrocyclopenta[a]phenanthren-17-yl] propanoat
Molekylvægt (g/mol) 347.51
Formel vægt 347,254 g/mol
Procent renhed 98 atom % D, min 98% kemisk renhed
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.