Learn More
ITS Europe™ Ultra-High Free Chlorine Test Strips
Product Code. 11732947
Change view
Click to view available options
Quantity:
50/Bottle
Unit Size:
Pack of 50
Product Code. Quantity Unit Size
11732947 50/Bottle Pack of 50
1 options
Product Code. 11732947 Supplier ITS Europe™ Supplier No. 480024
  • ONLINE EXCLUSIVE PRICE
    154.32 DKK
    132.61 DKK / Pakke med 50
    Spare 21.71 DKK
    14% Slukket
View all promotions
Forventet forsendelse: 14-12-2026
Patented technology provides a very high range of chlorine detection capacity in a convenient test strip. ITS Europe™ Ultra-High Free Chlorine Test Strips are supplied in a bottle of 50 strips.
  • Very high range of detection.
  • Detection range: 0 - 750ppm (mg/L).
  • Designed to resist interference from monochloramines
  • Ideal product when testing in a food processing environment.
  • Perfect for determining whether or not you have the correct amount of sanitizer present.
  • All reagents are impregnated on the test strip pad, and no external chemicals are needed to perform the water quality test.
  • Test time: 31 seconds.
Specifications
Analytes Chlorine
No. of Tests 50
Quantity 50/Bottle
Range 0 - 750ppm (mg/L)