Få mere at vide
Tesofensine, TRC
Artikelnummer. 30138760
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138760 10 mg
30003727 1 mg
30005635 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138760 Leverandør Toronto Research Chemicals Leverandørnr. TRCT13650010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 195875-84-4
Molekylær formel C17 H23 Cl2 N O
Molekylvægt (g/mol) 328.28
Synonym (1R,2R,3S,5S)-3-(3,4-Dichlorophenyl)-2-(ethoxymethyl)-8-methyl-8-azabicyclo[3.2.1]octane, [1R-(2-endo,3-exo)]-3-(3,4-dichlorophenyl)-2-(ethoxymethyl)-8-methyl-8-azabicyclo[3.2.1]octane, NS 2330;
IUPAC navn (1S,3S,4R,5R)-3-(3,4-dichlorophenyl)-4-(ethoxymethyl)-8-methyl-8-azabicyclo[3.2.1]octane
SMIL CCOC[C@H]1[C@H]2CC[C@@H](C[C@@H]1c3ccc(Cl)c(Cl)c3)N2C
Tekniske data
Kemisk navn eller materiale Tesofensine
Mængde 10 mg
InChI formel InChI=1S/C17H23Cl2NO/c1-3-21-10-14-13(9-12-5-7-17(14)20(12)2)11-4-6-15(18)16(19)8-11/h4,6,8,12-14,17H,3,5,7,9-10H2,1-2H3/t12-,13+,14+,17+/m0/s1
Formel vægt 327.1157