Få mere at vide
tert-Pentyl-d6 Chloride, TRC
Artikelnummer. 30418586
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30418586 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418586 Leverandør Toronto Research Chemicals Leverandørnr. TRCP27981750MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Pentyl-d6 Chloride
CAS 89603-08-7
Molekylær formel C5H5D6Cl
Mængde 50 mg
Synonym 1,1-Dimethylpropyl-d6 Chloride, 2-Chloro-2-methylbutane-d6, 2-Methyl-2-chlorobutane-d6, NSC 7900-d6, tert-Amyl Chloride-d6, 2-Chloro-2-methylbutane-d6
InChI formel InChI=1S/C5H11Cl/c1-4-5(2,3)6/h4H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])C(Cl)(CC)C([2H])([2H])[2H]
IUPAC navn 2-chloro-1,1,1-trideuterio-2-(trideuteriomethyl)butane
Molekylvægt (g/mol) 112.63
Formel vægt 112.0926