missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl propiolate, 98%, Thermo Scientific Chemicals

Catalog Number p-7033868
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
13831-03-3
C7H10O2
126.16
MFCD00060100
XGTPDIIFEPTULX-UHFFFAOYSA-N
tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
543038
tert-butyl prop-2-ynoate
CC(C)(C)OC(=O)C#C
This item is not returnable. View return policy
13831-03-3
C7H10O2
126.16
MFCD00060100
XGTPDIIFEPTULX-UHFFFAOYSA-N
tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
543038
tert-butyl prop-2-ynoate
CC(C)(C)OC(=O)C#C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 18°C to 20°C
Density 0.919
Boiling Point 52°C to 53°C (27 mmHg)
Flash Point 28°C (82°F)
Linear Formula HC≡CCO2C(CH3)3
Refractive Index 1.418
Quantity 1 g
UN Number UN3272
Beilstein 1747175
Sensitivity Air Sensitive
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material tert-Butyl propiolate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'