missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Thermo Scientific™

Catalog Number p-2705853
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
77152-97-7
C8H16N2O2S
204.288
MFCD02180883
OBDMXQCRRWGEQM-UHFFFAOYSA-N
tert-butyl n-3-amino-3-thioxopropyl carbamate, tert-butyl 3-amino-3-thioxopropylcarbamate, tert-butyl n-2-carbamothioylethyl carbamate, tert-butyl 3-amino-3-thioxopropyl carbamate, 3-tert-butoxycarbonyl amino propanethioamide, tert-butyl 3-amino-3-thioxoprop-1-yl carbamate, 2-thiocarbamoylethyl carbamic acid tert-butyl ester, carbamic acid,n-3-amino-3-thioxopropyl-, 1,1-dimethylethyl ester, tert-butyl 2-thiocarbamoylethylcarbamate, 3-tert-butoxycarbonylamino propanethioamide
2735653
tert-butyl N-(3-amino-3-sulfanylidenepropyl)carbamate
CC(C)(C)OC(=O)NCCC(=S)N
This item is not returnable. View return policy
77152-97-7
C8H16N2O2S
204.288
MFCD02180883
OBDMXQCRRWGEQM-UHFFFAOYSA-N
tert-butyl n-3-amino-3-thioxopropyl carbamate, tert-butyl 3-amino-3-thioxopropylcarbamate, tert-butyl n-2-carbamothioylethyl carbamate, tert-butyl 3-amino-3-thioxopropyl carbamate, 3-tert-butoxycarbonyl amino propanethioamide, tert-butyl 3-amino-3-thioxoprop-1-yl carbamate, 2-thiocarbamoylethyl carbamic acid tert-butyl ester, carbamic acid,n-3-amino-3-thioxopropyl-, 1,1-dimethylethyl ester, tert-butyl 2-thiocarbamoylethylcarbamate, 3-tert-butoxycarbonylamino propanethioamide
2735653
tert-butyl N-(3-amino-3-sulfanylidenepropyl)carbamate
CC(C)(C)OC(=O)NCCC(=S)N
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3COC(O)NH(CH2)2C(S)NH2
Quantity 1 g
Formula Weight 204.29
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(3-amino-3-thioxopropyl)carbamate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'