Få mere at vide
tert-Butyl Methylcarbamate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | tert-Butyl Methylcarbamate |
| CAS | 16066-84-5 |
| Molekylær formel | C6H13NO2 |
| Mængde | 1 g |
| Synonym | (tert-Butoxycarbonyl)methylamine, Methylcarbamic Acid tert-butyl Ester, N-(tert-Butoxycarbonyl)methylamine, N-Methylcarbamic Acid tert-Butyl Ester, tert-Butyl N-methylcarbamate, Methyl Carbamic Acid tert-Butyl Ester, Methyl Carbamic Acid 1,1-Dimethylethyl Ester, N-Methylcarbamic Acid 1,1-Dimethylethyl Ester |
| InChI formel | InChI=1S/C6H13NO2/c1-6(2,3)9-5(8)7-4/h1-4H3,(H,7,8) |
| SMIL | CNC(=O)OC(C)(C)C |
| IUPAC navn | tert-butyl N-methylcarbamate |
| Molekylvægt (g/mol) | 131.17 |
| Formel vægt | 131.0946 |