Få mere at vide
tert-Butyl Methyl Ether-d12, TRC
Artikelnummer. 30455051
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30455051 10 mg
30452931 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30455051 Leverandør Toronto Research Chemicals Leverandørnr. TRCT69337210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-tert-Butyl Ether D12
CAS 284487-64-5
Molekylær formel C5D12O
Mængde 10 mg
Synonym 1,1,1,3,3,3-Hexadeuterio-2-(trideuteriomethoxy)-2-(trideuteriomethyl)propane, tert-Butyl methyl ether-d12
InChI formel InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i1D3,2D3,3D3,4D3
SMIL [2H]C([2H])([2H])OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-(methoxy-d3)-2-(methyl-d3)propane-1,1,1,3,3,3-d6
Molekylvægt (g/mol) 100.22
Formel vægt 100.16