missing translation for 'onlineSavingsMsg'
Få mere at vide

tert-Butyl-d9 Methyl Ether, TRC

Artikelnummer. 30453698
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30453698 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453698 Leverandør Toronto Research Chemicals Leverandørnr. TRCT6933732.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Methyl-tert-Butyl Ether D9
CAS 1219795-06-8
Molekylær formel C5H3D9O
Mængde 2.5 mg
Synonym Methyl tert-Butyl Ether D9, 1,1,1,3,3,3-Hexadeuterio-2-methoxy-2-(trideuteriomethyl)propane, tert-Butyl methyl ether-d9, tert-Butyl-d9 methyl ether, Methyl-tert-butyl ether D9, Methyl tert-butyl-D9 ether, Methyl tert-butyl-d9 ether
InChI formel InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i1D3,2D3,3D3
SMIL COC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-methoxy-2-(methyl-d3)propane-1,1,1,3,3,3-d6
Molekylvægt (g/mol) 97.2
Formel vægt 97.15
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.