Få mere at vide
tert-Butyl-d9 Methyl Ether, TRC
Artikelnummer. 30451722
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30451722 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30451722 Leverandør Toronto Research Chemicals Leverandørnr. TRCT6933735MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Methyl-tert-Butyl Ether D9
CAS 1219795-06-8
Molekylær formel C5H3D9O
Mængde 5 mg
Synonym Methyl tert-Butyl Ether D9, 1,1,1,3,3,3-Hexadeuterio-2-methoxy-2-(trideuteriomethyl)propane, tert-Butyl methyl ether-d9, tert-Butyl-d9 methyl ether, Methyl-tert-butyl ether D9, Methyl tert-butyl-D9 ether, Methyl tert-butyl-d9 ether
InChI formel InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i1D3,2D3,3D3
SMIL COC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-methoxy-2-(methyl-d3)propane-1,1,1,3,3,3-d6
Molekylvægt (g/mol) 97.2
Formel vægt 97.15