Få mere at vide
tert-Butyl-d9-amine, TRC
Artikelnummer. 30346399
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30346399 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30346399 Leverandør Toronto Research Chemicals Leverandørnr. TRCB6907771G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Butyl-d9-amine
CAS 6045-08-5
Molekylær formel C4 D9 H2 N
Mængde 1 g
Synonym 2-Propan-1,1,1,3,3,3-d6-amine, 2-(methyl-d3)- (9CI), tert-Butyl-d9-amine (7CI, 8CI), 1,1,1,3,3,3-Hexadeuterio-2-(trideuteriomethyl)propan-2-amine, D9-tert-Butylamine, tert-Butylamine-D9
InChI formel InChI=1S/C4H11N/c1-4(2,3)5/h5H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(N)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-amine
Molekylvægt (g/mol) 82.1923
Formel vægt 82.1456