missing translation for 'onlineSavingsMsg'
Få mere at vide

tert-Butyl Bicyclo[2.2.2]phosphorothionate, TRC

Product Code. 30336949
Change view
Click to view available options
Mængde:
2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30336949 2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336949 Supplier Toronto Research Chemicals Supplier No. TRCB6943152MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale tert-Butyl Bicyclo[2.2.2]phosphorothionate
CAS 70636-86-1
Molekylær formel C8H15O3PS
Mængde 2 mg
Synonym 4-tert-Butyl-2,6,7-trioxa-1-phosphabicyclo[2.2.2]octane-1-thione, TBPS, tert-Butylbicyclophosphorothionate
InChI formel InChI=1S/C8H15O3PS/c1-7(2,3)8-4-9-12(13,10-5-8)11-6-8/h4-6H2,1-3H3
SMIL CC(C)(C)C12COP(=S)(OC1)OC2
IUPAC navn 4-tert-butyl-1-sulfanylidene-2,6,7-trioxa-1λ5-phosphabicyclo[2.2.2]octane
Molekylvægt (g/mol) 222.24
Formel vægt 222.048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.