Få mere at vide
Tert-butyl Alcohol-OD, TRC
Artikelnummer. 30427195
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30427195 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30427195 Leverandør Toronto Research Chemicals Leverandørnr. TRCT117542100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Tert-butyl Alcohol-OD
CAS 3972-25-6
Molekylær formel C4 D H9 O
Mængde 100 mg
Synonym 2-Propanol-d','2-methyl- (9CI), tert-Butyl alcohol-d (6CI','7CI','8CI), tert-Butyl hydroxide-d, tert-Butyl alcohol-OD, tert-Butanol-OD, 2-Methyl-2-propanol-d
InChI formel InChI=1S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i5D
SMIL [2H]OC(C)(C)C
IUPAC navn 2-deuteriooxy-2-methylpropane
Molekylvægt (g/mol) 75.128
Formel vægt 75.079