missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl acrylate, 99%, stab. with 15ppm 4-methoxyphenol, Thermo Scientific Chemicals

Catalog Number p-7047005
Click to view available options
Quantity:
100 mL
500 mL
1000 mL
1663-39-4
C7H12O2
128.17
MFCD00008809
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
15458
tert-butyl prop-2-enoate
CC(C)(C)OC(=O)C=C
1663-39-4
C7H12O2
128.17
MFCD00008809
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
15458
tert-butyl prop-2-enoate
CC(C)(C)OC(=O)C=C

tert-Butyl acrylate is a monomer used in the production of poly(tert-butyl acrylate) polymers, poly(acrylic acid) polyelectrolyte brushes and 3-(3-methyl-but-3-enyloxy)-propionic acid tert-butyl ester. It is also used in paints, coatings, high polymer paper conditioner, nursing polymer products and carbonyl reforming polyolefin. Further, it serves as an important raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acrylate is a monomer used in the production of poly(tert-butyl acrylate) polymers, poly(acrylic acid) polyelectrolyte brushes and 3-(3-methyl-but-3-enyloxy)-propionic acid tert-butyl ester. It is also used in paints, coatings, high polymer paper conditioner, nursing polymer products and carbonyl reforming polyolefin. Further, it serves as an important raw material in organic synthesis.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼-69°C
Density 0.883
Boiling Point 121°C
Flash Point 11°C (52°F)
Linear Formula CH2=CHCOOC(CH3)3
Refractive Index 1.411
Quantity 100 mL
UN Number UN3272
Beilstein 1742329
Solubility Information Slightly miscible with water.
Formula Weight 128.17
Percent Purity 99%
Chemical Name or Material tert-Butyl acrylate, stabilized with 15ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'