missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate, ≥97%, Thermo Scientific™

Catalog Number p-3111143
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
869901-02-0
C14H20N2O3S
296.385
MFCD06658981
XLGKMJFDRZHAEV-UHFFFAOYSA-N
7172298
tert-butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)C=O
This item is not returnable. View return policy
869901-02-0
C14H20N2O3S
296.385
MFCD06658981
XLGKMJFDRZHAEV-UHFFFAOYSA-N
7172298
tert-butyl 4-(4-formyl-1,3-thiazol-2-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)C2=NC(=CS2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym tert-butyl 4-4-formylthiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-formyl-1,3-thiazol-2-yl piperidine-1-carboxylate, tert-butyl 4-4-formyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-4-formylthiazol-2-yl piperidine-1-ca, tert-butyl4-4-formylthiazol-2-yl piperidine-1-carboxylate, 1,1-dimethylethyl 4-4-formyl-2-thiazolyl-1-piperidinecarboxylate, 4-4-formyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 1-piperidinecarboxylicacid, 4-4-formyl-2-thiazolyl-, 1,1-dimethylethyl ester, 4-4-formyl-2-thiazolyl-1-piperidinecarboxylic acid,1,1-dimethylethyl ester
Formula Weight 296.39
Percent Purity 97%
Chemical Name or Material tert-Butyl 4-(4-formyl-1, 3-thiazol-2-yl)tetrahydro-1(2H)-pyridinecarboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'