Få mere at vide
tert-Butyl-1,1,1,3,3,3-d6 Alcohol, CDN
Artikelnummer. 30321685
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30321685 1 g
30311646 0.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321685 Leverandør CDN Leverandørnr. CDND54481G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale tert-Butanol-D6 (dimethyl-D6)
CAS 53853-65-9
Molekylær formel C4 D6 H4 O
Mængde 1 g
Synonym 2-Propan-1,1,1,3,3,3-d6-ol, 2-methyl- (9 CI), 2-Methyl-2-propanol-1,1,1,3,3,3-d6, tert-Butyl-2,2,2,2',2',2'-d6 alcohol, tert-Butyl-1,1,1,3,3,3-d6 alcohol, tert-Butanol-d6, tert-Butanol-D6 (dimethyl-D6)
InChI formel InChI=1 S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])C(C)(O)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,3-hexadeuterio-2-methylpropan-2-ol
Molekylvægt (g/mol) 80.159
Formel vægt 80.111 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity