missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Terpineol, mixed isomers, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
2.5 L
Description
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
Solubility
Miscible with organic solvents. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Terpineol is used in perfumes, cosmetics and flavorings. It is also used in the chemical industries and personal-domestic use. It acts as a cleaning and washing agent, disinfectant, food additive and odor agent. It finds applications in fragrance compositions, disinfectants, polishes and various household cleaning products. It is also used as high-grade solvent and deodorizer.
Solubility
Miscible with organic solvents. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids.
Specifications
Melting Point | 28°C to 36°C |
Density | 0.934 |
Boiling Point | 213°C to 218°C |
Flash Point | 91°C (196°F) |
Refractive Index | 1.484 |
Quantity | 500 mL |
Solubility Information | Miscible with organic solvents. Slightly miscible with water. |
Formula Weight | 154.25 |
Percent Purity | 98% |
Chemical Name or Material | Terpineol, mixed isomers |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'