missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Terpin Hydrate, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Terpin Hydrate |
| CAS | 2451-01-6 |
| Molekylær formel | C10 H20 O2 . H2O |
| Mængde | 1 g |
| Synonym | cis-4-Hydroxy-alpha,alpha,4-trimethylcyclohexanemethanol monohydrate, p-Menthane-1,8-diol monohydrate, Terpin hydrate, Terpin monohydrate, Terpinene hydrate, Terpinol hydrate, cis-Terpin hydrate, cis-p-Methane-1,8-diol monohydrate, cis-p-Menthane-1,8-diol monohydrate |
| InChI formel | InChI=1S/C10H20O2.H2O/c1-9(2,11)8-4-6-10(3,12)7-5-8;/h8,11-12H,4-7H2,1-3H3;1H2/t8-,10+; |
| SMIL | O.CC(C)(O)[C@@H]1CC[C@](C)(O)CC1 |
| IUPAC navn | 4-(2-hydroxypropan-2-yl)-1-methylcyclohexan-1-ol;hydrate |
| Molekylvægt (g/mol) | 190.28 |
| Formel vægt | 190.1569 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?