Få mere at vide
Teroxalene Hydrochloride, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Teroxalene Hydrochloride |
| CAS | 3845-22-5 |
| Analyt- eller komponentnavne | Teroxalene Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C28 H41 Cl N2 O . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Piperazine, 1-(3-chloro-4-methylphenyl)-4-[6-[4-(1,1-dimethylpropyl)phenoxy]hexyl]-, hydrochloride (1:1), Piperazine, 1-(3-chloro-4-methylphenyl)-4-[6-[4-(1,1-dimethylpropyl)phenoxy]hexyl]-, monohydrochloride (9CI), Piperazine, 1-(3-chloro-p-tolyl)-4-[6-(p-tert-pentylphenoxy)hexyl]-, monohydrochloride (8CI), A 16612, Teroxalene hydrochloride |
| InChI formel | InChI=1S/C28H41ClN2O.ClH/c1-5-28(3,4)24-11-14-26(15-12-24)32-21-9-7-6-8-16-30-17-19-31(20-18-30)25-13-10-23(2)27(29)22-25;/h10-15,22H,5-9,16-21H2,1-4H3;1H |
| Løsningstype | Neat |
| SMIL | Cl.CCC(C)(C)c1ccc(OCCCCCCN2CCN(CC2)c3ccc(C)c(Cl)c3)cc1 |
| Vis mere |