Learn More
VACUUBRAND™ Terminal Inlet 2 Valve
74.65 DKK - 91.05 DKK
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11910162
|
VACUUBRAND™
20636746 |
Valve Head EX, double, 180° |
81.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
11920162
|
VACUUBRAND™
20636747 |
Valve Head EX, double, 90° |
74.65 DKK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
11930102
|
VACUUBRAND™
20636029 |
Valve Head IN, double, 180° |
91.05 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11930162
|
VACUUBRAND™
20636748 |
Valve, 90° |
84.70 DKK
Each |
In Stock
Log in to see stock. |
||||||
Description
Specifications
1 Each | |
Valve |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
VACUUBRAND™ Terminal Inlet 2 Valve