Learn More
Terfenadine, TRC
Product Code. 17972685
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972685 50 mg 50mg
17951365 5 g 5g
17912125 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17972685 Supplier Toronto Research Chemicals Supplier No. TRCT11450050MG
 Restricted Item
Forventet forsendelse: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50679-08-8
Molecular Formula C32 H41 N O2
Molecular Weight (g/mol) 471.67
Synonym Terfenadine, 1-Piperidinebutanol, α-[4-(1,1-dimethylethyl)phenyl]-4-(hydroxydiphenylmethyl)-, α-[4-(1,1-Dimethylethyl)phenyl]-4-(hydroxydiphenylmethyl)-1-piperidinebutanol, (±)-Terfenadine, 1-(4-tert-Butylphenyl)-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butan-1-ol, Allerplus, Cyater, Histadin, Histafen, MDL 9918, NSC 665802, Nebralin, Racemic terfenadine, Seldane, Teldane, Teldanex, Terdin, Terfenadine, Terfex, Terfinax, Ternadin, Triludan, dl-Terfenadine, α-(p-tert-Butylphenyl)-4-(α-hydroxy-α-phenylbenzyl)-1-piperidinebutanol
IUPAC Name 1-(4-tert-butylphenyl)-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butan-1-ol
SMILES CC(C)(C)c1ccc(cc1)C(O)CCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C32H41NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,34-35H,10,15,20-24H2,1-3H3
Formula Weight 471.3137
Chemical Name or Material Terfenadine