Få mere at vide
Terfenadine EP Impurity H, Mikromol™
Artikelnummer. 30049235
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049235 100 mg
30052220 25 mg
30460003 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049235 Leverandør Mikromol Leverandørnr. MM0055.10
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Terfenadine EP Impurity H
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (1-(4-(4-(1,1-Dimethylethyl)-phenyl)butyl)piperidin-4-yl)diphenylmethanol
CAS 147158-67-6
Analyt- eller komponentnavne (1-(4-(4-(1,1-Dimethylethyl)-phenyl)butyl)piperidin-4-yl)diphenylmethanol
Forsendelsestilstand Room Temperature
Molekylær formel C32 H41 N O
Certificeringer/compliance ISO 17025
Synonym [1-[4-[4-(1,1-Dimethylethyl)-phenyl]butyl]piperidin-4-yl]diphenylmethanol, Terfenadine Imp. H (EP), Terfenadine Impurity H
InChI formel InChI=1S/C32H41NO/c1-31(2,3)27-19-17-26(18-20-27)12-10-11-23-33-24-21-30(22-25-33)32(34,28-13-6-4-7-14-28)29-15-8-5-9-16-29/h4-9,13-20,30,34H,10-12,21-25H2,1-3H3
SMIL CC(C)(C)c1ccc(CCCCN2CCC(CC2)C(O)(c3ccccc3)c4ccccc4)cc1
IUPAC navn [1-[4-(4-tert-butylphenyl)butyl]piperidin-4-yl]-diphenylmethanol
Molekylvægt (g/mol) 455.67
Mængde 100 mg
Formel vægt 455.3188 g/mol