Få mere at vide
Terfenadine-d3, TRC
Artikelnummer. 30422815
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30422815 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422815 Leverandør Toronto Research Chemicals Leverandørnr. TRCT11450210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terfenadine-d3
CAS 192584-82-0
Molekylær formel C32 D3 H38 N O2
Mængde 10 mg
InChI formel InChI=1S/C32H41NO2/c1-31(2,3)26-18-16-25(17-19-26)30(34)15-10-22-33-23-20-29(21-24-33)32(35,27-11-6-4-7-12-27)28-13-8-5-9-14-28/h4-9,11-14,16-19,29-30,34-35H,10,15,20-24H2,1-3H3/i15D2,30D
SMIL [2H]C([2H])(CCN1CCC(CC1)C(O)(c2ccccc2)c3ccccc3)C([2H])(O)c4ccc(cc4)C(C)(C)C
IUPAC navn 1-(4-tert-butylphenyl)-1,2,2-trideuterio-4-[4-[hydroxy(diphenyl)methyl]piperidin-1-yl]butan-1-ol
Molekylvægt (g/mol) 474.692
Formel vægt 474.333