Learn More
Abnova™ TERC/CEN3q (Texas Red/FITC) FISH Probe Product Code.: 16053215
Abnova™ TERC/CEN3q (Texas Red/FITC) FISH Probe
Product Code. 16053215
100 μL, 100µL
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. 16053215
Brand: Abnova™ FG0139.100uL
Includes
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Content And Storage Store at 4°C in the dark.
Description Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Label or Dye FITC, Texas Red
For Use With (Application) Fluorescent In Situ Hybridization (Cell), Fluorescent In Situ Hybridization (Formalin/PFA-fixed paraffin-embedded sections)
Includes DAPI Counterstain (1500 ng/mL ) 250 uL
Location 3q26/3q12.1
Probe Approx. 230kb/500kb
Product Type TERC/CEN3q FISH Probe
Quantity 100 μL
Source Genomic DNA
Species Human
Show More Show Less
Product Content Correction
Product Title
Abnova™ TERC/CEN3q (Texas Red/FITC) FISH Probe >