Få mere at vide
Terbutryn-d5 (ethyl-d5), TRC
Artikelnummer. 30412817
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30412817 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412817 Leverandør Toronto Research Chemicals Leverandørnr. TRCT1099515MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terbutryn D5
CAS 1219804-47-3
Molekylær formel C10 2H5 H14 N5 S
Mængde 5 mg
Synonym Terbutryn D5 (ethyl D5), N'-tert-Butyl-6-methylsulfanyl-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
InChI formel InChI=1S/C10H19N5S/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15)/i1D3,6D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Nc1nc(NC(C)(C)C)nc(SC)n1
IUPAC navn 4-N-tert-butyl-6-methylsulfanyl-2-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 246.39
Formel vægt 246.1675