Få mere at vide
Terbutryn-d5 (ethyl-d5), CDN
Artikelnummer. 30323765
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323765 0.005 g
30322268 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323765 Leverandør CDN Leverandørnr. CDND71080.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terbutryn D5
CAS 1219804-47-3
Molekylær formel C10 2H5 H14 N5 S
Mængde 0.005 g
Synonym Terbutryn D5 (ethyl D5)
InChI formel InChI=1 S/C10H19N5S/c1-6-11-7-12-8(15-10(2,3)4)14-9(13-7)16-5/h6H2,1-5H3,(H2,11,12,13,14,15)/i1D3,6D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])Nc1nc(NC(C)(C)C)nc(SC)n1
IUPAC navn 4-N-tert-butyl-6-methylsulfanyl-2-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 246.39
Formel vægt 246.1675 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity