Få mere at vide
Terbuthylazine-(ethyl-d5), TRC
Artikelnummer. 30411717
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30411717 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30411717 Leverandør Toronto Research Chemicals Leverandørnr. TRCT1105045MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terbuthylazine D5
CAS 222986-60-9
Molekylær formel C9 2H5 H11 Cl N5
Mængde 5 mg
Synonym 1,3,5-Triazine-2,4-diamine','6-chloro-N-(1,1-dimethylethyl)-N'-(ethyl-d5)- (9CI), 6-Chloro-N-(1,1-dimethylethyl)-N'-(ethyl-d5)-1,3,5-triazine-2,4-diamine, 6-chloro-N2-(1,1-dimethylethyl)-N4-(ethyl-d5)-1,3,5-triazine-2,4-diamine, Terbuthylazine-d5, Terbuthylazine-D5 (N-ethyl-D5), Terbuthylazine D5 (ethyl D5)
InChI formel InChI=1S/C9H16ClN5/c1-5-11-7-12-6(10)13-8(14-7)15-9(2,3)4/h5H2,1-4H3,(H2,11,12,13,14,15)/i1D3,5D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Nc1nc(Cl)nc(NC(C)(C)C)n1
IUPAC navn 2-N-tert-butyl-6-chloro-4-N-(1,1,2,2,2-pentadeuterioethyl)-1,3,5-triazine-2,4-diamine
Molekylvægt (g/mol) 234.74
Formel vægt 234.1408