Få mere at vide
Terbutaline Sulfate, Mikromol™
Beskrivelse
Terbutaline Sulfate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Terbutaline Sulfate |
| CAS | 23031-32-5 |
| Analyt- eller komponentnavne | Terbutaline Sulfate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C12 H19 N O3 . H2 O4 S |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Terbutaline sulfate, Bis[(1RS)-1-(3,5-dihydroxyphenyl)-2-[(1,1-dimethylethyl)amino]ethanol] sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1), 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1) (salt) (9CI), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt) (8CI), (±)-Terbutaline hemisulfate, (±)-Terbutaline sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, (±)-, sulfate (2:1) (salt), 2-(tert-Butylamino)-1-(3,5-dihydroxyphenyl)ethanol sulfate (2:1), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt), (±)-, Brethaire, Brethine, Bricanyl, Brithine, Butaliret, Monovent, Terbasmin, Terbul, Terbutalin sulfate, Terbutaline hemisulfate, Terbutaline hemisulphate, dl-Terbutaline sulfate, Bambuterol Hydrochloride Impurity A as Sulphate, Bambuterol Impurity A as Sulphate |
| InChI formel | InChI=1S/2C12H19NO3.H2O4S/c2*1-12(2,3)13-7-11(16)8-4-9(14)6-10(15)5-8;1-5(2,3)4/h2*4-6,11,13-16H,7H2,1-3H3;(H2,1,2,3,4) |
| SMIL | CC(C)(C)NCC(O)c1cc(O)cc(O)c1.CC(C)(C)NCC(O)c2cc(O)cc(O)c2.OS(=O)(=O)O |
| IUPAC navn | 5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol;sulfuric acid |
| Vis mere |