missing translation for 'onlineSavingsMsg'
Få mere at vide

Terbutaline Sulfate, LoGiCal

Artikelnummer. 30464057
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30464057 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30464057 Leverandør LoGiCal Leverandørnr. LGCFOR0708.00

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Terbutaline Sulfate
CAS 23031-32-5
Analyt- eller komponentnavne Terbutaline Sulfate
Forsendelsestilstand Room Temperature
Molekylær formel 2 C12 H19 N O3 . H2 O4 S
Certificeringer/compliance ISO 9001
Synonym Terbutaline sulfate, Bis[(1RS)-1-(3,5-dihydroxyphenyl)-2-[(1,1-dimethylethyl)amino]ethanol] sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1), 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1) (salt) (9CI), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt) (8CI), (±)-Terbutaline hemisulfate, (±)-Terbutaline sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, (±)-, sulfate (2:1) (salt), 2-(tert-Butylamino)-1-(3,5-dihydroxyphenyl)ethanol sulfate (2:1), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt), (±)-, Brethaire, Brethine, Bricanyl, Brithine, Butaliret, Monovent, Terbasmin, Terbul, Terbutalin sulfate, Terbutaline hemisulfate, Terbutaline hemisulphate, dl-Terbutaline sulfate, Bambuterol Hydrochloride Impurity A as Sulphate, Bambuterol Impurity A as Sulphate
InChI formel InChI=1S/2C12H19NO3.H2O4S/c2*1-12(2,3)13-7-11(16)8-4-9(14)6-10(15)5-8;1-5(2,3)4/h2*4-6,11,13-16H,7H2,1-3H3;(H2,1,2,3,4)
Løsningstype Neat
SMIL CC(C)(C)NCC(O)c1cc(O)cc(O)c1.CC(C)(C)NCC(O)c2cc(O)cc(O)c2.OS(=O)(=O)O
IUPAC navn 5-[2-(tert-butylamino)-1-hydroxyethyl]benzene-1,3-diol;sulfuric acid
Industry Type Clinical Testing
Molekylvægt (g/mol) 548.65
Mængde 10 mg
Formel vægt 548.2404 g/mol
Vis mere Vis mindre
Produkttitel
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.