missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Terbutaline sulfate 100 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Terbutaline Sulfate |
| CAS | 23031-32-5 |
| Analyt- eller komponentnavne | Terbutaline Sulfate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C12 H19 N O3 . H2 O4 S |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Terbutaline sulfate, Bis[(1RS)-1-(3,5-dihydroxyphenyl)-2-[(1,1-dimethylethyl)amino]ethanol] sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1), 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, sulfate (2:1) (salt) (9CI), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt) (8CI), (±)-Terbutaline hemisulfate, (±)-Terbutaline sulfate, 1,3-Benzenediol, 5-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-, (±)-, sulfate (2:1) (salt), 2-(tert-Butylamino)-1-(3,5-dihydroxyphenyl)ethanol sulfate (2:1), Benzyl alcohol, α-[(tert-butylamino)methyl]-3,5-dihydroxy-, sulfate (2:1) (salt), (±)-, Brethaire, Brethine, Bricanyl, Brithine, Butaliret, Monovent, Terbasmin, Terbul, Terbutalin sulfate, Terbutaline hemisulfate, Terbutaline hemisulphate, dl-Terbutaline sulfate, Bambuterol Hydrochloride Impurity A as Sulphate, Bambuterol Impurity A as Sulphate |
| InChI formel | InChI=1S/2C12H19NO3.H2O4S/c2*1-12(2,3)13-7-11(16)8-4-9(14)6-10(15)5-8;1-5(2,3)4/h2*4-6,11,13-16H,7H2,1-3H3;(H2,1,2,3,4) |
| SMIL | CC(C)(C)NCC(O)c1cc(O)cc(O)c1.CC(C)(C)NCC(O)c2cc(O)cc(O)c2.OS(=O)(=O)O |
| Vis mere |
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?