Få mere at vide
Terbutaline-d9 3,5-Dibenzyl Ether, TRC
Artikelnummer. 30428618
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30428618 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30428618 Leverandør Toronto Research Chemicals Leverandørnr. TRCT10976250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terbutaline-d9 3,5-Dibenzyl Ether
CAS 1189428-81-6
Molekylær formel C26H22D9NO3
Mængde 50 mg
Synonym 1-[3,5-Bis(benzyloxy)phenyl]-2-(tert-butyl-d9-amino)ethanol, alpha-[[(1,1-Dimethylethyl-d9)amino]methyl]-3,5-bis(phenylmethoxy)bnzenemethanol
InChI formel InChI=1S/C26H31NO3/c1-26(2,3)27-17-25(28)22-14-23(29-18-20-10-6-4-7-11-20)16-24(15-22)30-19-21-12-8-5-9-13-21/h4-16,25,27-28H,17-19H2,1-3H3/i1D3,2D3,3D3
SMIL [2H]C([2H])([2H])C(NCC(O)c1cc(OCc2ccccc2)cc(OCc3ccccc3)c1)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 1-[3,5-bis(phenylmethoxy)phenyl]-2-[[1,1,1,3,3,3-hexadeuterio-2-(trideuteriomethyl)propan-2-yl]amino]ethanol
Molekylvægt (g/mol) 414.58
Formel vægt 414.2869