Få mere at vide
Terbinafine EP Impurity B (as Hydrochloride), Mikromol™
Beskrivelse
Terbinafine EP Impurity B (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | cis-Terbinafine Hydrochloride |
| CAS | 176168-78-8 |
| Analyt- eller komponentnavne | cis-Terbinafine Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C21 H25 N . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | (2Z)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride (cis-Terbinafine Hydrochloride), 1-Naphthalenemethanamine, N-[(2Z)-6,6-dimethyl-2-hepten-4-yn-1-yl]-N-methyl-, hydrochloride (1:1), 1-Naphthalenemethanamine, N-(6,6-dimethyl-2-hepten-4-ynyl)-N-methyl-, hydrochloride, (Z)-, 1-Naphthalenemethanamine, N-[(2Z)-6,6-dimethyl-2-hepten-4-ynyl]-N-methyl-, hydrochloride (9CI), (2Z)-N,6,6-Trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride, Terbinafine Hydrochloride Imp. B (EP) as Hydrochloride, cis-Terbinafine Hydrochloride, Terbinafine Related Compound B as Hydrochloride, Terbinafine Hydrochloride Impurity B as Hydrochloride, Terbinafine Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C21H25N.ClH/c1-21(2,3)15-8-5-9-16-22(4)17-19-13-10-12-18-11-6-7-14-20(18)19;/h5-7,9-14H,16-17H2,1-4H3;1H/b9-5-; |
| SMIL | Cl.CN(C\C=C/C#CC(C)(C)C)Cc1cccc2ccccc12 |
| IUPAC navn | (Z)-N,6,6-trimethyl-N-(naphthalen-1-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride |
| Vis mere |