Få mere at vide
Terbinafine-d7 Hydrochloride, TRC
Artikelnummer. 30422519
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30422519 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30422519 Leverandør Toronto Research Chemicals Leverandørnr. TRCT10750210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Terbinafine-d7 Hydrochloride
CAS 1185240-27-0
Renhedsgrad noter HPLC
Molekylær formel C21 H18 D7 N . H Cl
Mængde 10 mg
Synonym N-[(2,3,4,5,6,7,8-Heptadeuterionaphthalen-1-yl)methyl]-N,6,6-trimethylhept-2-en-4-yn-1-amine hydrochloride, N-(6,6-Dimethyl-2-hepten-4-yn-1-yl)-N-methyl-1-naphthalene-2,3,4,5,6,7,8-d7-methanamine hydrochloride, (EZ)-Terbinafine-d7 hydrochloride, (EZ)-Terbinafine-D7 (naphthalene-D7) hydrochloride
InChI formel InChI=1S/C21H25N.ClH/c1-21(2,3)15-8-5-9-16-22(4)17-19-13-10-12-18-11-6-7-14-20(18)19;/h5-7,9-14H,16-17H2,1-4H3;1H/b9-5+;/i6D,7D,10D,11D,12D,13D,14D;
SMIL Cl.[2H]c1c([2H])c([2H])c2c(CN(C)C\C=C\C#CC(C)(C)C)c([2H])c([2H])c([2H])c2c1[2H]
IUPAC navn (E)-N-[(2,3,4,5,6,7,8-heptadeuterionaphthalen-1-yl)methyl]-N,6,6-trimethylhept-2-en-4-yn-1-amine;hydrochloride
Molekylvægt (g/mol) 334.93
Formel vægt 334.2193
Procent renhed >95