Learn More
TentaGel™ MB-Br, Thermo Scientific Chemicals
Catalog Number 10031981
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
MFCD00217898
O-(2-Bromoethyl)polyethylene glycol polymer bound
MFCD00217898
O-(2-Bromoethyl)polyethylene glycol polymer bound
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesis
TRUSTED_SUSTAINABILITY
Color White to Yellow
Physical Form Powder
Loading 0.4 to 0.6mmol/g
Packaging Glass bottle
Particle Size 140 to 170 Micron
Quantity 5 g
Chemical Name or Material TentaGel™ MB-Br