Få mere at vide
Tenofovir Monohydrate, TRC
Artikelnummer. 30423369
Skift visning
Mængde:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423369 100 mg
30410296 1 g
30428103 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30423369 Leverandør Toronto Research Chemicals Leverandørnr. TRCT185533100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 206184-49-8
Molekylær formel C9 H14 N5 O4 P . H2 O
Molekylvægt (g/mol) 305.23
Synonym Phosphonic acid, P-[[(1R)-2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, hydrate (1:1), Phosphonic acid, [[(1R)-2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, monohydrate (9CI), Phosphonic acid, [[2-(6-amino-9H-purin-9-yl)-1-methylethoxy]methyl]-, monohydrate, (R)-, Tenofovir monohydrate, P-[[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic acid monohydrate, (R)-9-(2-Phosphonomethoxypropyl)adenine monohydrate, ([[(2R)-1-(6-Amino-9H-purin-9-yl)propan-2-yl]oxy]methyl)phosphonic acid monohydrate
IUPAC navn [(2R)-1-(6-aminopurin-9-yl)propan-2-yl]oxymethylphosphonic acid;hydrate
SMIL O.C[C@H](Cn1cnc2c(N)ncnc12)OCP(=O)(O)O
Tekniske data
Kemisk navn eller materiale Tenofovir Monohydrate
Mængde 100 mg
InChI formel InChI=1S/C9H14N5O4P.H2O/c1-6(18-5-19(15,16)17)2-14-4-13-7-8(10)11-3-12-9(7)14;/h3-4,6H,2,5H2,1H3,(H2,10,11,12)(H2,15,16,17);1H2/t6-;/m1./s1
Formel vægt 305.0889