Få mere at vide
Tenofovir Disoproxil Isopropoxycarbonyl, TRC
Artikelnummer. 17993685
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17993685 5 mg
17914525 1 mg
17960665 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17993685 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0185205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1244022-54-5
Molekylær formel C23 H36 N5 O12 P
Molekylvægt (g/mol) 605.53
Synonym 2,4,6,8-Tetraoxa-5-phosphanonanedioic acid, 5-[[(1R)-1-methyl-2-[6-[[(1-methylethoxy)carbonyl]amino]-9H-purin-9-yl]ethoxy]methyl]-, 1,9-bis(1-methylethyl) ester, 5-oxide, [[(2R)-1-[6-(Isopropyloxycarbonylamino)-9H-purin-9-yl]propan-2-yloxy]methyl(isopropyloxycarbonyloxymethoxy)phosphoryl]oxymethyl Isopropyl Carbonate
IUPAC navn propan-2-yl [[(2R)-1-[6-(propan-2-yloxycarbonylamino)purin-9-yl]propan-2-yl]oxymethyl-(propan-2-yloxycarbonyloxymethoxy)phosphoryl]oxymethyl carbonate
SMIL CC(C)OC(=O)Nc1ncnc2c1ncn2C[C@@H](C)OCP(=O)(OCOC(=O)OC(C)C)OCOC(=O)OC(C)C
Tekniske data
Kemisk navn eller materiale Tenofovir Disoproxil Isopropoxycarbonyl
Mængde 5 mg
InChI formel InChI=1S/C23H36N5O12P/c1-14(2)38-21(29)27-19-18-20(25-9-24-19)28(10-26-18)8-17(7)35-13-41(32,36-11-33-22(30)39-15(3)4)37-12-34-23(31)40-16(5)6/h9-10,14-17H,8,11-13H2,1-7H3,(H,24,25,27,29)/t17-/m1/s1
Formel vægt 605.2098