missing translation for 'onlineSavingsMsg'
Få mere at vide

Tenofovir Diphosphate Triethylamine Salt (Mixture of Diastereomers), TRC

Artikelnummer. 30417500
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417500 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30417500 Leverandør Toronto Research Chemicals Leverandørnr. TRCT0185250.5MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Tenofovir diphosphate triethylamine salt
CAS 2122333-63-3
Renhedsgrad noter HPLC
Molekylær formel C9 H16 N5 O10 P3 . C6 H15 N
Mængde 0.5 mg
Synonym Diphosphoric Acid Triethylamine Salt ','Anhydride with [[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid Triethylamine Salt, PMPApp Triethylamine Salt
InChI formel InChI=1S/C9H16N5O10P3.C6H15N/c1-6(2-14-4-13-7-8(10)11-3-12-9(7)14)22-5-25(15,16)23-27(20,21)24-26(17,18)19;1-4-7(5-2)6-3/h3-4,6H,2,5H2,1H3,(H,15,16)(H,20,21)(H2,10,11,12)(H2,17,18,19);4-6H2,1-3H3/t6-;/m1./s1
SMIL CCN(CC)CC.C[C@H](Cn1cnc2c(N)ncnc12)OCP(=O)(O)OP(=O)(O)OP(=O)(O)O
IUPAC navn [(1R)-2-(6-aminopurin-9-yl)-1-methyl-ethoxy]methyl-[hydroxy(phosphonooxy)phosphoryl]oxy-phosphinic acid;x-triethylamine
Molekylvægt (g/mol) 447.17
Formel vægt 447.011
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.