Få mere at vide
Tenofovir Diphosphate Triethylamine Salt (mixture of diastereomers), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Tenofovir diphosphate triethylamine salt |
| CAS | 2122333-63-3 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C9 H16 N5 O10 P3 . C6 H15 N |
| Mængde | 10 mg |
| Synonym | Diphosphoric Acid Triethylamine Salt ','Anhydride with [[(1R)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]phosphonic Acid Triethylamine Salt, PMPApp Triethylamine Salt |
| InChI formel | InChI=1S/C9H16N5O10P3.C6H15N/c1-6(2-14-4-13-7-8(10)11-3-12-9(7)14)22-5-25(15,16)23-27(20,21)24-26(17,18)19;1-4-7(5-2)6-3/h3-4,6H,2,5H2,1H3,(H,15,16)(H,20,21)(H2,10,11,12)(H2,17,18,19);4-6H2,1-3H3/t6-;/m1./s1 |
| SMIL | CCN(CC)CC.C[C@H](Cn1cnc2c(N)ncnc12)OCP(=O)(O)OP(=O)(O)OP(=O)(O)O |
| IUPAC navn | [(1R)-2-(6-aminopurin-9-yl)-1-methyl-ethoxy]methyl-[hydroxy(phosphonooxy)phosphoryl]oxy-phosphinic acid;x-triethylamine |
| Molekylvægt (g/mol) | 447.17 |
| Vis mere |