Learn More
Memmert™ Temperature Profile Write/Read Unit (Pc/Chip Card) Up To 40 Ramps, For Ctc256
Memmert™ Temperature Profile Write/Read Unit for programming via PC allows writing and reading from the chip card.
Brand:  Memmert™ CTC256E05284
Product Code. 16384767
  • 945.00 DKK / Each
Only null left
Estimated Shipment: 18-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • For programming via PC
  • For writing to and reading from the chip card
  • Up to 40 ramps
Specifications
Specifications
Up to 40 ramps, programming via PC
Models 200, 400, 500, 108, 153, 246, 256
Temperature Profile Unit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Temperature Profile Write/Read Unit (Pc/Chip Card) Up To 40 Ramps, For Ctc256 >