Learn More
BINDER™ Temperature and Humidity Calibration Certificate
Calibration certificate including 27 temperature measurements and 1 humidity measurement.
Brand: BINDER™ 8012-2577
This item is not returnable.
View return policy
Description
- 27 temperature measuring points
- 1 humidity measuring point, at 25°C/60% RH, or at specified values
Specifications
KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600 | |
DIN12880 | |
1 Day | |
Calibration Certificate |
Product Content Correction
Product Title
BINDER™ Temperature and Humidity Calibration Certificate > For use with KBF 130, KBF 260, KBF 470, KBF 720, KBF 1060, KBF 1600
Spot an opportunity for improvement?Share a Content Correction