Learn More
BINDER™ Temperature Analog Output
For Use With: ED 56, ED 115, ED 260, ED 720
Brand:  BINDER™ 8012-1623
Product Code. 16404339
  • 9040.00 DKK / Each
Only null left
Estimated Shipment: 28-08-2025
Description
  • Output not adjustable
  • 4 to 20 mA
Specifications
Specifications
Analog output 4-20 mA - for temperature values (output not adjustable)
Plastic
Temperature
BINDER Chambers ED 56, ED 115, ED 260, ED 720
Analog Output
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
BINDER™ Temperature Analog Output > For Use With: ED 56, ED 115, ED 260, ED 720