Learn More
BINDER™ Temperature Analog Output
For Use With: ED 56, ED 115, ED 260, ED 720
Brand: BINDER™ 8012-1623
This item is not returnable.
View return policy
Description
- Output not adjustable
- 4 to 20 mA
Specifications
Analog output 4-20 mA - for temperature values (output not adjustable) | |
Plastic | |
Temperature |
BINDER Chambers ED 56, ED 115, ED 260, ED 720 | |
Analog Output |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Temperature Analog Output > For Use With: ED 56, ED 115, ED 260, ED 720
Spot an opportunity for improvement?Share a Content Correction