Få mere at vide
Teicoplanin A2 (Mixture of 2-1, 2-2, 2-3, 2-4 and 2-5), TRC
Artikelnummer. 30010517
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010517 250 mg
30004585 50 mg
30008028 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30010517 Leverandør Toronto Research Chemicals Leverandørnr. TRCT015500250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 61036-64-4
Tekniske data
Kemisk navn eller materiale Teicoplanin A2 (Mixture of 2-1, 2-2, 2-3, 2-4 and 2-5)
Mængde 250 mg
Synonym Teicoplanin A2, Teichomycin A2, MDL-507, Targocid, Targosid;